Cefoxitin sodium 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
45528-0010
45528-0050
TS45528-0010EA
138
USD
TS45528-0010
TS45528-0050
Cefoxitin sodium 94%
Cefoxitin sodium
Formula:
C₁₆H₁₆N₃NaO₇S₂ MW: 449.428 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00079042 CAS Number: 33564-30-6 |
Specification Test Results
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
HPLC | ≥93 % |
Specific optical rotation | +206° to +214° (20°C, 589 nm) (c= 1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...