Valacyclovir hydrochloride hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
L-Valine 2-(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-ylmethoxy)ethyl Ester Hydrochloride
46226-0050
46226-0250
TS46226-0050EA
148
USD
TS46226-0050
TS46226-0250
Valacyclovir hydrochloride hydrate
Valacyclovir hydrochloride hydrate
Formula:
C₁₃H₂₀N₆O₄ MW: 360.80 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01861507 CAS Number: 124832-27-5 |
Specification Test Results
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Water | 5.0 to 11.0 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...