Pirfenidone

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

46472-0010 46472-2500
TS46472-0010EA 354 USD
TS46472-0010 TS46472-2500
Pirfenidone
Pirfenidone
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Melting Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00866047
CAS Number: 53179-13-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 111 °C
Loss on drying ≤0.5 % (1 g, 80 °C, 4 hr, 75 kPa)
Solubility (5% in CH2Cl2) Clear colorless to yellow
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR