Maltitol 95%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol

29580-0010 29580-0250 29580-2500
TS29580-0010EA 691 USD
TS29580-0010 TS29580-0250 TS29580-2500
Maltitol 95%
Maltitol
Formula: C₁₂H₂₄O₁₁
MW: 344.31 g/mol
Melting Pt: 147…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00006600
CAS Number: 585-88-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Water ≤1 %
Solubility (10 % in water) Clear colorless
HPLC ≥94 % (ELSD)
Specific optical rotation +105.5° to +108.5° (20 °C, 589 nm) (c=5, H2O) (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR