Fisetin
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,7,3',4'-Tetrahydroxyflavone
11915-0010
119150050
TS11915-0010EA
168
USD
TS11915-0010
TS119150050
Fisetin
Fisetin
Formula:
C₁₅H₁₀O₆ MW: 286.24 g/mol Melting Pt: >330 °C |
MDL Number:
MFCD00006829 CAS Number: 528-48-3 EINECS: 208-434-4 |
Specification Test Results
Appearance | White to amber to dark green Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤5 % |
HPLC | ≥90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...