Finasteride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide

45800-0010 45800-0050
TS45800-0010EA 236 USD
TS45800-0010 TS45800-0050
Finasteride 98%
Finasteride
Formula: C₂₃H₃₆N₂O₂
MW: 372.55 g/mol
Melting Pt: 252…254 °C
Storage Temperature: Freezer
MDL Number: MFCD00869737
CAS Number: 98319-26-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR