Fenofibrate 98%

Supplier: Thermo Scientific Chemicals
Warning

46178-1000 46178-0050 46178-0250
TS46178-1000EA 486 USD
TS46178-1000 TS46178-0050 TS46178-0250
Fenofibrate 98%
Fenofibrate
Formula: C₂₀H₂₁ClO₄
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00133314
CAS Number: 49562-28-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
Loss on drying ≤0.5 % (60 °C) (vacuum)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR