D-Biotin-N-succinimidyl ester 98%
Supplier: Thermo Scientific Chemicals
43930-0010
43930-2500
TS43930-0010EA
662
USD
TS43930-0010
TS43930-2500
D-Biotin-N-succinimidyl ester 98%
D-Biotin-N-succinimidyl ester
Formula:
C₁₄H₁₉N₃O₅S Melting Pt: 202.3…202.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD00078531 CAS Number: 35013-72-0 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤2 % |
Elemental Analysis | 12.00 to 12.62 % N |
Specific optical rotation | +52° to +54° (20°C, 589 nm) (c=1 in DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...