Boc-b-Alanine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-beta-alanine
, Boc-β-Ala-OH
, BOC-beta-alanine
43765-0250
43765-0050
43765-1000
TS43765-0250EA
81.13
USD
TS43765-0250
TS43765-1000
TS43765-0050
Boc-b-Alanine 99%
Boc-b-Alanine
Formula:
C₈H₁₅NO₄ MW: 189.2 g/mol Melting Pt: 77.5…78.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037291 CAS Number: 3303-84-2 |
Specification Test Results
Appearance | White Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 76 to 80 °C |
Elemental Analysis | 50.02 to 51.54 % C |
Elemental Analysis | 7.29 to 7.51 % N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...