D-(+)-Cellobiose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

10846-1000 10846-0050 10846-0250 10846-5000
TS10846-1000EA 238 USD
TS10846-1000 TS10846-0050 TS10846-0250 TS10846-5000
D-(+)-Cellobiose 98%
D(+)-Cellobiose
Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 239 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Solubility (5 % in water) Clear colorless
HPLC ≥97.5 % (ELSD)
Specific optical rotation +33° to +35° (20°C, 589 nm) (c=1, water) (after 15 h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR