(±)-Aspartic acid 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Aminobutanedioic acid, DL-Aspartic acid

10503-0010 10503-0050 10503-2500
TS10503-0010EA 102 USD
TS10503-0010 TS10503-0050 TS10503-2500
(±)-Aspartic acid 99+%
(±)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 210-513-3

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 h)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, 2 N HCl, 430 nm) 1 cm cell
Specific optical rotation -0.3° to +0.3° (20°C, 589 nm) (c=10, 2 N HCl)
Titration with NaOH 99.0 to 101.0 %
Ammonium (NH4) ≤500 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR