Actinomycin D 93%

Supplier: Thermo Scientific Chemicals
Danger

29494-0010 29494-0050 29494-0250 29494-1000
TS29494-0010EA 60 USD
TS29494-0010 TS29494-0050 TS29494-0250 TS29494-1000
Actinomycin D 93%
Actinomycin D
Formula: C₆₂H₈₆N₁₂O₁₆
Melting Pt: 245…252 °C
Storage Temperature: Freezer
MDL Number: MFCD00005033
CAS Number: 50-76-0
EINECS: 200-063-6
UN: 3462
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Red Crystalline powder
Infrared spectrum Conforms
HPLC ≥91 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR