Tetracycline hydrochloride can be used as secondary standard
Supplier: Thermo Scientific Chemicals
|
Warning
|
23310-0025
23310-5000
TS23310-0025EA
353
USD
TS23310-0025
TS23310-5000
Tetracycline hydrochloride can be used as secondary standard
Tetracycline hydrochloride
Formula:
C₂₂H₂₅ClN₂O₈ MW: 480.9 g/mol Melting Pt: 223 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD00078142 CAS Number: 64-75-5 EINECS: 200-593-8 |
Specification Test Results
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Purity | ≥80 % (as is, calculated) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...