N,N-Dimethylglycine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
34520-0050
34520-0250
TS34520-0050EA
81
USD
TS34520-0050
TS34520-0250
N,N-Dimethylglycine 97%
N,N-Dimethylglycine
Formula:
C₄H₉NO₂ Melting Pt: 175…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004283 CAS Number: 1118-68-9 EINECS: 214-267-8 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 175 to 181 °C |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...