D-Cycloserine 98%
Supplier: Thermo Scientific Chemicals
22848-0050
22848-0250
22848-0010
TS22848-0050EA
293
USD
TS22848-0050
TS22848-0250
TS22848-0010
D-Cycloserine 98%
D-Cycloserine
Formula:
C₃H₆N₂O₂ Melting Pt: 147 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005353 CAS Number: 68-41-7 EINECS: 200-688-4 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (60°C, 3 hrs) (vacuum) |
UV | at 285 nm A: ≤0.8 |
Condensation products | (0.04 % in 0.1 N NaOH) |
Potency | ≥900 µg/mg |
Residue after ignition | ≤0.5 % |
pH | 5.5 to 6.5 (10 % solution) |
Specific optical rotation | +108° to +114° (20°C, 589 nm) (c=5, 2N NaOH) |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...