L(-)-Sorbose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

22596-0010 22596-1000 22596-2500
TS22596-0010EA 1519 USD
TS22596-0010 TS22596-1000 TS22596-2500
L(-)-Sorbose 98%
L(-)-Sorbose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 380
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00151097
CAS Number: 87-79-6
EINECS: 201-771-8
Merck Index: 12,08874

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 % (ELSD)
Specific optical rotation -42° to -44° (20°C, 589 nm) (c=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR