Cytosine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol

16176-0050 16176-0250 16176-1000
TS16176-0050EA 102 USD
TS16176-0050 TS16176-0250 TS16176-1000
Cytosine 99+%
Cytosine
Formula: C₄H₅N₃O
MW: 111.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 % (800°C)
HPLC ≥99.0 %
UV spectrophotometry molar absorptivity 10400 +/- 2%, on dry basis
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR