Saponin (from Quillaja Saponaria Molina) pract.

Supplier: Thermo Scientific Chemicals

41923-0025 41923-5000
TS41923-0025EA 1333 USD
TS41923-0025 TS41923-1000 TS41923-5000
Saponin (from Quillaja Saponaria Molina) pract.
Saponin
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2

Order Now

Specification Test Results

Appearance Dark red to tan Powder
Infrared spectrum Conforms
Solubility (20 % in water) Clear to hazy foaming solution
Moisture ≤10 %
Ash ≤10 %
pH 3.8 to 4.6 (20 % solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR