Toluene-α-sulfonyl fluoride 99%

Supplier: Thermo Scientific Chemicals
Danger

21574-0010 21574-0050 21574-0100 21574-0500
TS21574-0010EA 64 USD
TS21574-0010 TS21574-0050 TS21574-0100 TS21574-0500
Toluene-α-sulfonyl fluoride 99%
Toluene-α-sulphonyl fluoride
Formula: C₇H₇FO₂S
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White Needles and/or agglomerates
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR