L(+)-Histidine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
16615-0250
16615-1000
16615-5000
TS16615-0250EA
44
USD
TS16615-0250
TS16615-1000
TS16615-5000
L(+)-Histidine 98%
L(+)-Histidine
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol Melting Pt: 282 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064315 CAS Number: 71-00-1 EINECS: 200-745-3 Merck Index: 13,04741 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.2 % (105°C, 3 h) |
Sulfated ash | ≤0.1 % |
UV transmittance | ≥98 % (c=5, H2O, 430 nm) 1 cm cell |
pH | 7.0 to 8.5 (2 % in water) |
Specific optical rotation | +12.0° to +14.0° (20°C, 589 nm) (c=11, 6 N HCl) (on dry substance) |
Titration with HClO4 | 97.5 to 102.5 % (On dry substance) |
Chloride (Cl) | ≤200 ppm |
Sulfate (SO4) | ≤200 ppm |
Ammonium (NH4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...