L(+)-Glutamic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

15621-0010 15621-0050 15621-1000 15621-2500
TS15621-0010EA 85 USD
TS15621-0010 TS15621-0050 TS15621-1000 TS15621-2500
L(+)-Glutamic acid 99%
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Boiling Pt: 325
Melting Pt: 160 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
Specific optical rotation +30.5° to +32.5° (20°C, 589 nm) (c=10, 2N HCl) (on dried substance)
Titration with NaOH 98.5 to 100.5 % (On dried substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤30 ppm
Arsenic oxide (As2O3) ≤1 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids not detectable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR