4-Pentenoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
26902-0050
26902-0250
26902-1000
TS26902-0050EA
34
USD
TS26902-0050
TS26902-0250
TS26902-1000
4-Pentenoic acid 99%
4-Pentenoic acid
Formula:
C₅H₈O₂ Boiling Pt: 83…84 °C (12 mmHg) Melting Pt: –22.5 °C Density: 0.975 g/cm³ Flash Pt: 89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004408 CAS Number: 591-80-0 EINECS: 209-732-7 UN: 3265 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4273 to 1.4303 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific gravity | (25°C) 0.971 to 0.979 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...