2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,8,9-Tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, Triisobutylphosphatrane, 2,8,9-Tri-i-butyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, [Nitrilotrisethylenetris(isobutylimino)]phosphine

42825-0010 42825-0050
TS42825-0010EA 104 USD
TS42825-0010 TS42825-0050
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane 97%
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Formula: C₁₈H₃₉N₄P
MW: 342.51 g/mol
Boiling Pt: 110…118 °C (.5 mmHg)
Density: 0.964 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD03840347
CAS Number: 331465-71-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Elemental Analysis 15.70 to 17.01 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR