6-Carboxyfluorescein 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM
42580-1000
42580-5000
TS42580-1000EA
136
USD
TS42580-1000
TS42580-5000
6-Carboxyfluorescein 96%
6-Carboxyfluorescein
Formula:
C₂₁H₁₂O₇ MW: 376.32 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00036873 CAS Number: 3301-79-9 |
Specification Test Results
Appearance | Yellow to orange Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...