Nitromethane 96% ACS

Supplier: Thermo Scientific Chemicals
Warning

42401-0010 42401-0025 42401-5000
TS42401-0010EA 150 USD
TS42401-0010 TS42401-0025 TS42401-5000
Nitromethane 96% ACS
Nitromethane
Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 101.2
Melting Pt: -29
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3 II
Merck Index: 13,06644

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3805 to 1.3825 (20°C, 589 nm)
Water ≤0.05 % (Coulometric)
GC ≥95 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR