Trimethyloxonium tetrafluoroborate 95%

Supplier: Thermo Scientific Chemicals
Danger

42196-0025 42196-0100
TS42196-0025EA 68 USD
TS42196-0025 TS42196-0100
Trimethyloxonium tetrafluoroborate 95%
Trimethyloxonium tetrafluoroborate
Formula: C₃H₉BF₄O
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011798
CAS Number: 420-37-1
EINECS: 206-994-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Titration with NaOH 94.0 to 106.0 %
Refractive index 1.4420 to 1.4450 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR