Nigrosine, water soluble, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Acid Black 2

41568-1000 41568-5000
TS41568-1000EA 71 USD
TS41568-1000 TS41568-5000
Nigrosine, water soluble, pure
Nigrosine
Storage Temperature: Ambient
MDL Number: MFCD00044681
CAS Number: 8005-03-6

Order Now

Specification Test Results

Appearance Dark red to black Crystalline powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR