Crystal violet, certified
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
40583-0250
40583-1000
40583-5000
TS40583-0250EA
39
USD
TS40583-0250
TS40583-1000
TS40583-5000
Crystal violet, certified
Crystal Violet
Formula:
C₂₅H₃₀ClN₃ MW: 407.99 g/mol Melting Pt: 173 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9 III Merck Index: 13,04406 |
Specification Test Results
Appearance | Dark green Crystalline powder |
Infrared spectrum | Conforms |
Dye content | ≥88 % |
UV Lambda max | 589 to 594 nm |
Absorption Ratio P-15/P+15 | 0.98 to 1.20 |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...