Bromophenol blue ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

40314-0050 40314-0100 40314-0500
TS40314-0050EA 30 USD
TS40314-0050 TS40314-0100 TS40314-0500
Bromophenol blue ACS
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 279 °C
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now

Specification Test Results

Appearance Pink-orange to red to purple Powder
Infrared spectrum Conforms
Visual transition interval from pH 3.0 (yellow) to pH 4.6 (blue)
Clarity of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR