Trimethyl orthopropionate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
38462-0250
38462-1000
TS38462-0250EA
53
USD
TS38462-0250
TS38462-1000
Trimethyl orthopropionate 97%
Trimethyl orthopropionate
Formula:
C₆H₁₄O₃ Boiling Pt: 121…122 °C (760 mmHg) Density: 0.944 g/cm³ Flash Pt: 19 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043305 CAS Number: 24823-81-2 UN: 3271 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3970 to 1.3990 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...