Tetraoctylammonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetra-n-octylammonium bromide
35210-0050
35210-0250
35210-1000
TS35210-0050EA
33
USD
TS35210-0050
TS35210-0250
TS35210-1000
Tetraoctylammonium bromide 98%
Tetraoctylammonium bromide
Formula:
C₃₂H₆₈BrN Melting Pt: 95…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011863 CAS Number: 14866-33-2 EINECS: 238-936-9 |
Specification Test Results
Appearance | White to cream Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 95 to 99 °C |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...