Brucine, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3-Dimethoxystrychnidin-10-one, 10,11-Dimethoxystrychnine

27872-0050 27872-0500 27872-1000 27872-5000
TS27872-0050EA 31 USD
TS27872-0050 TS27872-0500 TS27872-1000 TS27872-5000
Brucine, anhydrous 99%
Brucine
Formula: C₂₃H₂₆N₂O₄
MW: 394.47 g/mol
Melting Pt: 174…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00005942
CAS Number: 357-57-3
UN: 1570
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 179 °C (on dry substance)
Loss on drying ≤1 % (105 °C, 3 hrs)
Sulfated ash ≤0.1 %
Specific optical rotation -118° to -128° (20 °C, 589 nm) (c=2.5, CHCl3)
Titration with HClO4 ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR