Triphosgene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bis(trichloromethyl) carbonate

25895-0050 25895-0100 25895-0500 25895-2500
TS25895-0050EA 69 USD
TS25895-0050 TS25895-0100 TS25895-0500 TS25895-2500
Triphosgene 99%
Triphosgene
Formula: C₃Cl₆O₃
MW: 296.75 g/mol
Boiling Pt: 203…206 °C (760 mmHg)
Melting Pt: 77…81 °C
Density: 1.78 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00062848
CAS Number: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 77 to 81 °C
Assay ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR