m-Cresol purple, pure indicator grade

Supplier: Thermo Scientific Chemicals

Synonyms: m-Cresolsulfonphthalein, 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 2',2''-Dimethylphenolsulfonphthalein

22820-0050 22820-0100 22820-0500
TS22820-0050EA 61 USD
TS22820-0050 TS22820-0100 TS22820-0500
m-Cresol purple, pure indicator grade
m-Cresol purple
Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Solubility (0.1 % in 50 % ethanol) Clear
Specific extinction E (1%/1cm) ≥900 (578 nm, in buffer)
Visual transition interval from pH 7.4 (yellow) to pH 9.0 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR