Thiourea 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

22005-0010 22005-0050 22005-2500
TS22005-0010EA 96 USD
TS22005-0010 TS22005-0050 TS22005-2500
Thiourea 99+% for analysis
Thiourea
Formula: NH₂CSNH₂
MW: 76.12 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9 III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Titration Iodimetric ≥99.0 %
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm
Zinc (Zn) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR