Benzophenone 99+%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
21968-0010
21968-0050
21968-0500
21968-2500
TS21968-0010EA
98
USD
TS21968-0010
TS21968-0050
TS21968-0500
TS21968-2500
Benzophenone 99+%, pure
Benzophenone
Formula:
C₁₃H₁₀O Boiling Pt: 305.4 °C (1013 hPa) Melting Pt: 47…49 °C Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 Merck Index: 13,01099 |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 47 to 49 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...