Chloro(diisopropyl)phosphine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di-i-propylchlorophosphine

32155-0050 32155-0250
TS32155-0050EA 88 USD
TS32155-0050 TS32155-0250
Chloro(diisopropyl)phosphine 96%
Chloro(diisopropyl)phosphine
Formula: C₆H₁₄ClP
Boiling Pt: 69 °C (33 mmHg)
Density: 0.95 g/cm³
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00015027
CAS Number: 40244-90-4
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
Titration Argentometric ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR