Acridine orange (hemizinc salt) ∼55% (dye content), pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride
30091-0250
30091-1000
TS30091-0250EA
67
USD
TS30091-0250
TS30091-1000
Acridine orange (hemizinc salt) ∼55% (dye content), pure
Acridine orange (hemizinc salt)
Formula:
C₁₇H₂₀ClN₃·HCl·½ZnCl₂ MW: 369.96 g/mol Melting Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081043 CAS Number: 10127-02-3 EINECS: 233-353-6 UN: 3143 ADR: 6.1,III |
Specification Test Results
Appearance | Red-brown Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...