tert-Butyldichlorophosphine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: t-Butyldichlorophosphine

30053-0025 30053-0100
TS30053-0025EA 58 USD
TS30053-0025 TS30053-0100
tert-Butyldichlorophosphine
tert-Butyldichlorophosphine
Formula: C₄H₉Cl₂P
Boiling Pt: 142…150 °C (760 mmHg)
Melting Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00013615
CAS Number: 25979-07-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR