1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane

27582-0025 27582-5000
TS27582-0025EA 328 USD
TS27582-0025 TS27582-5000
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%
1,10-Diaza-18-crown-6 (1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane, Kryptofix® 22)
Formula: C₁₂H₂₆N₂O₄
MW: 262.35 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005112
CAS Number: 23978-55-4
EINECS: 245-965-0
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white Glistening crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR