Solvent Blue 38, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN
21217-0250
21217-1000
TS21217-0250EA
57
USD
TS21217-0250
TS21217-1000
Solvent Blue 38, pure
Solvent Blue 38
Formula:
C32H12CuN8Na2O6S2 MW: 778.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00071424 CAS Number: 1328-51-4 |
Specification Test Results
Appearance | Blue Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...