Eosin Y (yellowish), pure indicator grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
20193-0250
20193-1000
20193-2500
TS20193-0250EA
45
USD
TS20193-0250
TS20193-1000
TS20193-2500
Eosin Y (yellowish), pure indicator grade
Eosin Y (yellowish)
Formula:
C₂₀H₆Br₄Na₂O₅ Melting Pt: >300 °C (dec.) Density: 1.018 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 EINECS: 241-409-6 Merck Index: 1303634 |
Specification Test Results
Appearance | Dark red-brown Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Loss on drying | ≤10 % |
UV Lambda max | 511 to 519 nm |
Specific extinction E (1%/1cm) | ≥1100 (lambda max, 0.001M Na2CO3) on dried substance |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...