Hydrazinium sulphate 99+%

Supplier: Thermo Scientific Chemicals
Danger

20186-1000 20186-5000
TS20186-1000EA 71 USD
TS20186-1000 TS20186-5000
Hydrazinium sulphate 99+%
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
Melting Pt: 254 °C
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with NaOH ≥99.0 %
Total chloride ≤20 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Zinc (Zn) ≤5 ppm
Total silicon (Si) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR