α-Naphtholphthalein, pure indicator

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

19012-0010 19012-0050 19012-0250
TS19012-0010EA 49 USD
TS19012-0010 TS19012-0050 TS19012-0250
α-Naphtholphthalein, pure indicator
α-Naphtholphthalein
Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
Melting Pt: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00036202
CAS Number: 596-01-0
EINECS: 209-875-5

Order Now

Specification Test Results

Appearance Beige to pink to brown Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (110°C, 1 h)
UV Lambda max 648 to 654 nm (in 0.1N NaOH)
Specific absorbance A (1%/1cm) ≥550 (at lamda max)
Visual transition interval from pH 7.3 (pink-yellow) to pH 8.7 (green-blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR