Boranepyridine complex 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine Borane, Borane, pyridine complex

17701-0250 17701-1000
TS17701-0250EA 83 USD
TS17701-0250 TS17701-1000
Boranepyridine complex 95%
Boranepyridine complex
Formula: C₅H₈BN
Melting Pt: 10…11 °C
Density: 0.92 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012435
CAS Number: 110-51-0
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Titration ≥94 % (Active hydrogen)
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR