4-tert-Butylpyrocatechol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15435-0025
15435-0100
15435-1000
15435-5000
TS15435-0025EA
448
USD
TS15435-0025
TS15435-0100
TS15435-1000
TS15435-5000
4-tert-Butylpyrocatechol 99%
4-tert-Butylpyrocatechol
Formula:
C₁₀H₁₄O₂ Boiling Pt: 285 °C (760 mmHg) Melting Pt: 53…56 °C Density: 1.049 g/cm³ Flash Pt: 129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002201 CAS Number: 98-29-3 EINECS: 202-653-9 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to cream Flakes |
Infrared spectrum | Conforms |
Melting point | 53 to 56 °C |
Water | ≤0.3 % (K.F.) |
GC | ≤0.7 % Catechol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...