Neocuproine hemihydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,9-Dimethyl-1,10-phenanthroline hemihydrate

15332-0050 15332-0250
TS15332-0050EA 89 USD
TS15332-0050 TS15332-0250
Neocuproine hemihydrate 99+%
Neocuproine hemihydrate
Formula: C₂₈H₂₆N₄O
MW: 434.53 g/mol
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00149306
CAS Number: 34302-69-7
EINECS: 207-601-9
Merck Index: 1306477

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 159 to 164 °C
Titration with HClO4 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR