Acid red 27 85%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Amaranth
, Azo Rubine S
, Food Red No. 2
, Bordeaux S
, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate
, Wool Red 40 F
, Sodium 3-hydroxy-4-((4-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulfonate
, Naphthol Red
15303-1000
15303-5000
15303-0250
TS15303-1000EA
98
USD
TS15303-1000
TS15303-5000
TS15303-0250
Acid red 27 85%, pure
Acid red 27
Formula:
C₂₀H₁₄N₂O₁₀S₃ MW: 604.46 g/mol Melting Pt: 218…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004076 CAS Number: 915-67-3 |
Specification Test Results
Appearance | Dark red-brown to brown Crystalline powder |
Infrared spectrum | Conforms |
UV spectrophotometry | ≥80 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...