Triphenylphosphine oxide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14043-0250
14043-1000
14043-5000
TS14043-0250EA
30
USD
TS14043-0250
TS14043-1000
TS14043-5000
Triphenylphosphine oxide 99%
Triphenylphosphine oxide
Formula:
C₁₈H₁₅OP Boiling Pt: ∼360 °C (1013 hPa) Melting Pt: 154…158 °C Flash Pt: 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002080 CAS Number: 791-28-6 EINECS: 212-338-8 |
Specification Test Results
Appearance | White to pink-brown Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 154 to 158 °C |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...