p-Terphenyl 99+%, pure

Supplier: Thermo Scientific Chemicals
Warning

13792-0250 13792-1000 13792-5000
TS13792-0250EA 42 USD
TS13792-0250 TS13792-1000 TS13792-5000
p-Terphenyl 99+%, pure
p-Terphenyl
Formula: C₁₈H₁₄
Boiling Pt: 383 °C (760 mmHg)
Melting Pt: 209…216 °C
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00003061
CAS Number: 92-94-4
EINECS: 202-205-2

Order Now

Specification Test Results

Appearance White to light beige Glistening crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 211 to 216 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR